Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PREX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534960
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534960 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534960 Supplier Invitrogen™ Supplier No. PA534960
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: MCF-7, human PREX1-transfected 293T cells (partial). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene acts as a guanine nucleotide exchange factor for the RHO family of small GTP-binding proteins (RACs). It has been shown to bind to and activate RAC1 by exchanging bound GDP for free GTP. The encoded protein, which is found mainly in the cytoplasm, is activated by phosphatidylinositol-3,4,5-trisphosphate and the beta-gamma subunits of heterotrimeric G proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PREX1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene PREX1
Gene Accession No. Q8TCU6
Gene Alias G630042G04; KIAA1415; OTTHUMP00000031228; Phosphatidylinositol 3,4,5-trisphosphate-dependent Rac exchanger 1 protein; phosphatidylinositol-3,4,5-trisphosphate dependent Rac exchange factor 1; phosphatidylinositol-3,4,5-trisphosphate-dependent Rac exchange factor 1; PIP3 dependent Rac exchange factor 1; PREX 1; PREX1; P-Rex1; PREX-1; ptdIns(3,4,5)-dependent Rac exchanger 1; RGD1306534; SET domain containing 6; Setd6
Gene Symbols PREX1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PREX1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57580
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.