Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PREX1 Rabbit anti-Human, Mouse, Non-human Primate, Rat, Polyclonal, FabGennix

Catalog No. 502208720
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-8720 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-8720 Supplier Fabgennix Inc Supplier No. PREX101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PREX1 Polyclonal antibody specifically detects PREX1 in Human, Mouse, Non-human Primate, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen PREX1
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene PREX1
Gene Accession No. Q69ZK0, Q8TCU6
Gene Alias G630042G04; KIAA1415; OTTHUMP00000031228; Phosphatidylinositol 3,4,5-trisphosphate-dependent Rac exchanger 1 protein; phosphatidylinositol-3,4,5-trisphosphate dependent Rac exchange factor 1; phosphatidylinositol-3,4,5-trisphosphate-dependent Rac exchange factor 1; PIP3 dependent Rac exchange factor 1; PREX 1; PREX1; P-Rex1; PREX-1; ptdIns(3,4,5)-dependent Rac exchanger 1; RGD1306534; SET domain containing 6; Setd6
Host Species Rabbit
Immunogen Synthetic peptide corresponding to aino acid region 815-836 of Human PREX1. Peptide was covalently modified.Sequence:QEEDQADSAFPLLSLGPRLSLC
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 277360, 311647, 458315, 57580
Target Species Human, Mouse, Non-human Primate, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.