Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PREX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567400
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567400 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567400 Supplier Invitrogen™ Supplier No. PA567400
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HRLMKHDLKVV ENVIAKSLLI KSNEGSYGFG LEDKNKVPII KLVEKGSNAE MAGMEVGKKI FAINGDLVFM RPFNEVDCFL KSCLNSRKPL RVLVSTKP Highest antigen sequence identity to the following orthologs - mouse 98%, rat 98%.

Functions as a RAC1 guanine nucleotide exchange factor (GEF), activating Rac proteins by exchanging bound GDP for free GTP. Its activity is synergistically activated by phosphatidylinositol 3,4,5-trisphosphate and the beta gamma subunits of heterotrimeric G protein. Mediates the activation of RAC1 in a PI3K-dependent manner. May be an important mediator of Rac signaling, acting directly downstream of both G protein-coupled receptors and phosphoinositide 3-kinase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PREX2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PREX2
Gene Accession No. Q70Z35
Gene Alias 6230420N16Rik; AI316880; AI553603; C030045D06Rik; D430013K02; DEP domain containing 2; DEP domain-containing protein 2; DEP.2; Depdc2; phosphatidylinositol 3,4,5-trisphosphate-dependent Rac exchanger 2 protein; phosphatidylinositol-3,4,5-trisphosphate dependent Rac exchange factor 2; phosphatidylinositol-3,4,5-trisphosphate-dependent Rac exchange factor 2; PPP1R129; Prex2; P-Rex2; protein phosphatase 1, regulatory subunit 129; ptdIns(3,4,5)-dependent Rac exchanger 2; si:ch211-278g15.1
Gene Symbols PREX2
Host Species Rabbit
Immunogen Recombinant Human PREX2. Recombinant protein control fragment (Product #RP-108064).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80243
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.