Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRG2 Monoclonal Antibody (C3)
GREENER_CHOICE

Catalog No. PIMA541636
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541636 200 μL
PIMA543624 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA541636 Supplier Invitrogen™ Supplier No. MA541636
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PRG2 Monoclonal Antibody for Western Blot

The protein encoded by this gene is the predominant constituent of the crystalline core of the eosinophil granule. High levels of the proform of this protein are also present in placenta and pregnancy serum, where it exists as a complex with several other proteins including pregnancy-associated plasma protein A (PAPPA), angiotensinogen (AGT), and C3dg. This protein may be involved in antiparasitic defense mechanisms as a cytotoxin and helminthotoxin, and in immune hypersensitivity reactions. It is directly implicated in epithelial cell damage, exfoliation, and bronchospasm in allergic diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRG2
Applications Western Blot
Classification Monoclonal
Clone C3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene PRG2
Gene Accession No. Q63189
Gene Alias BMPG; bone marrow proteoglycan; bone-marrow proteoglycan; EMBP; Eosinophil granule major basic protein; eosinophil major basic protein; LPR3; major basic protein 1; MBP; MBP1; Mbp-1; mMBP; mMBP-1; natural killer cell activator; Pregnancy-associated major basic protein; PRG2; PRG-2; proMBP; Proteoglycan 2; proteoglycan 2 preproprotein; proteoglycan 2, bone marrow; proteoglycan 2, bone marrow (natural killer cell activator, eosinophil granule major basic protein); proteoglycan 2, pro eosinophil major basic protein
Gene Symbols PRG2
Host Species Mouse
Immunogen Recombinant protein Major Basic Protein. The antigen corresponds to amino acid range 80-219 of the target protein.
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58826
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.