Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRG3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551198
Description
The antibody detects endogenous levels of total PRG3 protein.
Possesses similar cytotoxic and cytostimulatory activities to PRG2/MBP. In vitro, stimulates neutrophil superoxide production and IL8 release, and histamine and leukotriene C4 release from basophils. [UniProt].
Specifications
PRG3 | |
Polyclonal | |
Unconjugated | |
PRG3 | |
eosinophil major basic protein 2; Eosinophil major basic protein homolog; major basic protein 2; MBP2; MBPH; Prepro-major basic protein homolog; prepro-MBPH; PRG3; proteoglycan 3; proteoglycan 3, pro eosinophil major basic protein 2; UNQ486/PRO1002 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10394 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.3 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9Y2Y8 | |
PRG3 | |
Full length fusion protein of human PRG3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction