Learn More
Invitrogen™ PRH1 Polyclonal Antibody

Description
Peptide sequence: VSQEDVPLVI SDGGDSEQFI DEERQGPPLG GQQSQPSAGD GNQNDGPQQG Sequence homology: Human: 100%.
Specifications
Specifications
| Antigen | PRH1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | PRH1 |
| Gene Accession No. | P02810 |
| Gene Alias | Db-F; Db-s; PA; Parotid acidic protein; parotid double-band protein; Parotid isoelectric focusing variant protein; parotid proline-rich protein 1/2; Peptide P-C; PIF-F; PIF-S; Pr1/Pr2; PRH1; PRH2; proline rich protein HaeIII subfamily 1; proline-rich protein HaeIII subfamily 1; Protein A; Protein C; PRP-1/PRP-2; PRP-3/PRP-4; Salivary acidic proline-rich phosphoprotein 1/2; Salivary acidic proline-rich phosphoprotein 3/4 |
| Gene Symbols | PRH1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.