Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRICKLE1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572728
Description
PRICKLE1 Polyclonal Antibody for Western Blot, ELISA
Involved in the planar cell polarity pathway that controls convergent extension during gastrulation and neural tube closure. Convergent extension is a complex morphogenetic process during which cells elongate, move mediolaterally, and intercalate between neighboring cells, leading to convergence toward the mediolateral axis and extension along the anteroposterior axis. Necessary for nuclear localization of REST. May serve as nuclear receptor.
Specifications
PRICKLE1 | |
Polyclonal | |
Unconjugated | |
PRICKLE1 | |
1110058P22Rik; AW215793; b2b019Clo; EPM1B; mpk1; Prickle; prickle homolog 1; prickle homolog 1 (Drosophila); prickle like 1; prickle planar cell polarity protein 1; PRICKLE1; prickle-like 1; prickle-like protein 1; REST (RE-1 silencing transcription factor)/NRSF (neuron-restrictive silencer factor)-interacting LIM domain protein; REST/NRSF interacting LIM domain protein; REST/NRSF-interacting LIM domain protein 1; Rilp | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
106042, 144165, 315259 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q3U5C7, Q71QF9, Q96MT3 | |
PRICKLE1 | |
PRICKLE1 antibody for a 19 amino acid synthetic peptide near the carboxy terminus of human PRICKLE1. The immunogen is within the last 50 amino acids of PRICKLE1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction