Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRICKLE1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 225891AP150UL

Description
Involved in the planar cell polarity pathway that controls convergent extension during gastrulation and neural tube closure. Convergent extension is a complex morphogenetic process during which cells elongate, move mediolaterally, and intercalate between neighboring cells, leading to convergence toward the mediolateral axis and extension along the anteroposterior axis. Necessary for nuclear localization of REST. May serve as nuclear receptor.Specifications
PRICKLE1 | |
Polyclonal | |
Unconjugated | |
PRICKLE1 | |
EPM1B, prickle homolog 1 (Drosophila), Prickle like protein 1, PRICKLE1, RILP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
106042, 144165 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.57 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3U5C7, Q96MT3 | |
Prickle1 | |
PRICKLE1 Fusion Protein Ag18407 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction