Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prickle1 Rabbit anti-Mouse, Unconjugated, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABN169925UG
Description
Anti-Prickle1 , Cat. No. ABN1699, is a highly specific rabbit polyclonal antibody that targets Prickle-like protein 1 and has been tested in Western Blotting.
Specifications
Prickle1 | |
Polyclonal | |
Unconjugated | |
Prickle1 | |
His-tagged Recombinant Protein corresponding to the C-terminus of Mouse Prickle-like protein 1. | |
Affinity Purified | |
RUO | |
Primary | |
Mouse | |
Antibodies | |
IgG |
Western Blot | |
Please refer to lot specific datasheet. | |
Purified rabbit polyclonal antibody in PBS with 0.05% sodium azide and 50% glycerol. | |
Rabbit | |
∽94 kDa observed; 94.13 kDa calculated. Uncharacterized bands may be observed in some lysate(s). | |
25 μg | |
Neuroscience | |
This rabbit polyclonal antibody detects Prickle-like protein 1 in murine species. | |
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction