Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ PRICKLE2, Mouse, Unlabeled, Clone: 3B4.1,

Catalog No. MABN1529MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABN1529MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1529MI Supplier MilliporeSigma™ Supplier No. MABN1529
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Prickle-like protein 2 (UniProt Q7Z3G6) is encoded by the PRICKLE2 (also known as EPM5) gene (Gene ID 166336) in human. Prickle2 is a nuclear membrane associated protein that belongs to a highly conserved Prickle, Espinas, Testin (PET) and Linl-1, Isl-1, and Mec-3 (LIM) domain-containing protein family. Prickle is part of the non-canonical Wnt signaling pathway that establishes planar cell polarity. It plays a critical role in neuron architecture and function. In mice it is shown to be highly expressed in the hippocampus region where it localizes to the post-synaptic density (PSD). It interacts the PSD-95 and NMDA receptor, the two proteins implicated in Autism spectrum disorders. Loss of Prickle2 is known to reduce number and size of miniature synaptic currents, indicating deficits in basal synaptic transmission. Mice deficient in Prickle2 exhibit lower seizure threshold and PRICKLE2 mutations in humans are associated with progressive myoclonic epilepsy type 5. Ref.: Sowers, LP., et al.. (2013), Mol. Psychiatry. 18, 1077-1089.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRICKLE2
Applications Western Blot
Classification Monoclonal
Clone 3B4.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG2aκ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene PRICKLE2, EPM5
Gene Accession No. Q7Z3G6
Host Species Mouse
Immunogen GST-tagged recombinant human PRICKLE2 internal fragment.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_942559
Test Specificity Clone 3B4.1 targets an internal PRICKLE2 epitope conserved between human and murine species.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.