Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRICKLE2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545653
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545653 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545653 Supplier Invitrogen™ Supplier No. PA545653
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: IPQPARLRYV TSDELLHKYS SYGLPKSSTL GGRGQLHSRK RQKSKNCIIS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.

This gene encodes a homolog of Drosophila prickle. The exact function of this gene is not known, however, studies in mice suggest that it may be involved in seizure prevention. Mutations in this gene are associated with progressive myoclonic epilepsy type 5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRICKLE2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PRICKLE2
Gene Accession No. Q7Z3G6
Gene Alias 6230400G14Rik; 6720451F06Rik; DKFZp686D143; DKFZp686H1748; DKFZp686M031; EPM5; EPM5p; mpk2; PLP2; PRIC2; prickle homolog 2; prickle homolog 2 (Drosophila); prickle planar cell polarity protein 2; PRICKLE2; prickle-like 2; prickle-like protein 2
Gene Symbols PRICKLE2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human PRICKLE2 (aa 795-844).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 166336
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.