Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRICKLE2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572729
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572729 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572729 Supplier Invitrogen™ Supplier No. PA572729
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PRICKLE2 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

This gene encodes a homolog of Drosophila prickle. The exact function of this gene is not known, however, studies in mice suggest that it may be involved in seizure prevention. Mutations in this gene are associated with progressive myoclonic epilepsy type 5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRICKLE2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene PRICKLE2
Gene Accession No. Q7Z3G6, Q80Y24
Gene Alias 6230400G14Rik; 6720451F06Rik; DKFZp686D143; DKFZp686H1748; DKFZp686M031; EPM5; EPM5p; mpk2; PLP2; PRIC2; prickle homolog 2; prickle homolog 2 (Drosophila); prickle planar cell polarity protein 2; PRICKLE2; prickle-like 2; prickle-like protein 2
Gene Symbols PRICKLE2
Host Species Rabbit
Immunogen PRICKLE2 antibody for a 15 amino acid synthetic peptide near the center of human PRICKLE2. The immunogen is within amino acids 420 - 470 of PRICKLE2.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 166336, 243548, 312563
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.