Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRICKLE2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576331
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576331 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576331 Supplier Invitrogen™ Supplier No. PA576331
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

This gene encodes a homolog of Drosophila prickle. The exact function of this gene is not known, however, studies in mice suggest that it may be involved in seizure prevention. Mutations in this gene are associated with progressive myoclonic epilepsy type 5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRICKLE2
Applications Immunohistochemistry, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PRICKLE2
Gene Accession No. Q7Z3G6, Q80Y24
Gene Alias 6230400G14Rik; 6720451F06Rik; DKFZp686D143; DKFZp686H1748; DKFZp686M031; EPM5; EPM5p; mpk2; PLP2; PRIC2; prickle homolog 2; prickle homolog 2 (Drosophila); prickle planar cell polarity protein 2; PRICKLE2; prickle-like 2; prickle-like protein 2
Gene Symbols PRICKLE2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Prickle 2: 401-500.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 166336, 243548
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.