Learn More
Invitrogen™ PRICKLE2 Polyclonal Antibody

Description
Immunogen sequence: QEMEGNLHQL SNPIGYRDLQ SHGRMHQSFD FDGGMAGSKL PGQEGVRIQP MSERTRRRAT SRDDNRRFRP HRSRRSRRSR SDNALHLASE REAISRLKDR PPLRAREDYD QFMRQRSFQE SMGHGSRRDL YGQCPRTVSD LALQNAFGDR WGPYFAEYDW CSTCSSSSES DNEGYFLGEP IPQPARLRYV TSDELLHKYS SYGLPKSSTL GGRGQLHSRK RQKSKNCIIS; Positive Samples: Mouse brain, Rat brain; Cellular Location: Nucleus membrane.
Specifications
Specifications
| Antigen | PRICKLE2 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.46 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | PRICKLE2 |
| Gene Accession No. | Q7Z3G6, Q80Y24 |
| Gene Alias | 6230400G14Rik; 6720451F06Rik; DKFZp686D143; DKFZp686H1748; DKFZp686M031; EPM5; EPM5p; mpk2; PLP2; PRIC2; prickle homolog 2; prickle homolog 2 (Drosophila); prickle planar cell polarity protein 2; PRICKLE2; prickle-like 2; prickle-like protein 2 |
| Gene Symbols | PRICKLE2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.