Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRICKLE3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551497
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551497 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551497 Supplier Invitrogen™ Supplier No. PA551497
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LIFCSRACSL GSEPTAPGPS RRSWSAGPVT APLAASTASF SAVKGASETT TKGTSTELAP ATGPEEPSRF LRGAPHRHSM PELGLRSVPE PPPESPGQPN LRPDDSAFGR QS Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 75%.

LIM domain only 6 is a three LIM domain-containing protein. The LIM domain is a cysteine-rich sequence motif that binds zinc atoms to form a specific protein-binding interface for protein-protein interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRICKLE3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Prickle3
Gene Accession No. O43900
Gene Alias LIM domain only 6; LIM domain only protein 6; LMO6; LMO-6; Pk3; prickle homolog 3; prickle homolog 3 (Drosophila); prickle planar cell polarity protein 3; Prickle3; Prickle-like protein 3; Sfc16; triple LIM domain protein 6
Gene Symbols Prickle3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRICKLE3. Recombinant protein control fragment (Product #RP-91981).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4007
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.