Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRICKLE3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564879
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564879 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564879 Supplier Invitrogen™ Supplier No. PA564879
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LIFCSRACSLG SEPTAPGPSR RSWSAGPVTA PLAASTASFS AVKGASETTT KGTSTELAPA TGPEEPSRFL RGAPHRHSMP ELGLRSVPEP PPESPGQPNL RPDDSAFGRQ S Highest antigen sequence identity to the following orthologs - mouse 79%, rat 75%.

LIM domain only 6 is a three LIM domain-containing protein. The LIM domain is a cysteine-rich sequence motif that binds zinc atoms to form a specific protein-binding interface for protein-protein interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRICKLE3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Prickle3
Gene Accession No. O43900
Gene Alias LIM domain only 6; LIM domain only protein 6; LMO6; LMO-6; Pk3; prickle homolog 3; prickle homolog 3 (Drosophila); prickle planar cell polarity protein 3; Prickle3; Prickle-like protein 3; Sfc16; triple LIM domain protein 6
Gene Symbols Prickle3
Host Species Rabbit
Immunogen Recombinant protein fragment of Human PRICKLE3 (Product #RP-91981)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4007
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.