Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRICKLE3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 190981AP150UL

Description
LIM domain only 6 is a three LIM domain-containing protein. The LIM domain is a cysteine-rich sequence motif that binds zinc atoms to form a specific protein-binding interface for protein-protein interactions.Specifications
PRICKLE3 | |
Polyclonal | |
Unconjugated | |
PRICKLE3 | |
LIM domain only protein 6, LMO 6, LMO6, prickle homolog 3 (Drosophila), Prickle like protein 3, PRICKLE3, Triple LIM domain protein 6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
317380, 4007, 54630 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43900, Q80VL3 | |
Prickle3 | |
PRICKLE3 Fusion Protein Ag13632 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction