Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRICKLE4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572730
Description
PRICKLE4 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
C6ORF49 is a member of the LIM domain protein family.
Specifications
PRICKLE4 | |
Polyclonal | |
Unconjugated | |
PRICKLE4 | |
C6orf49; OBTP; OEBT; overexpressed breast tumor protein; over-expressed breast tumor protein; overexpressed breast tumour protein; pric4; prickle homolog 4; prickle planar cell polarity protein 4; PRICKLE4; prickle-like protein 4; TOMM6; translocase of outer mitochondrial membrane 6 homolog | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29964 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q2TBC4 | |
PRICKLE4 | |
PRICKLE4 antibody for an 18 amino acid synthetic peptide near the amino terminus of human PRICKLE4. The immunogen is within the first 50 amino acids of PRICKLE4. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction