Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Pridopidine
GREENER_CHOICESDP

Catalog No. 763050 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-305-0 50 mg
76-301-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 76-305-0 Supplier Tocris Bioscience™ Supplier No. 7630/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Highly selective σ1 agonist

Pridopidine is a selective high affinity σ-1 receptor (S1R) agonist (Ki = 7.1 nM in both HEK293 cells and rat striatal membranes). Pridopidine displays ∼30-fold and ∼100-fold selectivity over the dopamine D3 receptor and the D2 receptor, respectively. Pridopidine is a low affinity dopamine D2 receptor antagonist (Ki (low) = 17.5 μM; Ki (high) = 7.5 μM). Pridopidine upregulates gene expression induced by BDNF in rat striatum in vivo in an S1R-dependent manner. Pridopidine improves motor function and shows neuroprotective effects in Huntington's disease animal models. Pridopidine restores neuromuscular junction activity in amyotrophic lateral sclerosis models in vitro and in vivo. Orally bioavailable.

Specifications

Chemical Name or Material 4-[3-(Methylsulfonyl)phenyl]-1-propylpiperidine
CAS 346688-38-8
Quantity 50 mg
Target Sigma1 Receptor Agonists
Molecular Formula C15H23NO2S
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.