Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRIM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116707
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116707 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116707 Supplier Invitrogen™ Supplier No. PA5116707
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse spleen Immunogen sequence: CCSSADICPK CWTLMTMAIR IIDRALKEDF GFKHRLWVYS GRRGVHCWVC DESVRKLSSA VRSGIVEYLS LVKGGQDVKK KVHLSEKIHP FIRKSINIIK KYFEEYALVN QDILENKESW DKILALVPET IHDELQQSFQ KSHNSLQRWE HLKKVASRYQ NNIKNDKYGP WLEWEIMLQY CFPRLDINVS KGINHLLKSP FSVHPKTGRI SVPIDLQKVD QFDPFTVPTI SFICRELDAI STNEEEKEEN EAESDVKHRT RDYKKTSLAP YVKVFEHFLE NLDKSRKGEL LKKSDLQKDF.

The replication of DNA in eukaryotic cells is carried out by a complex chromosomal replication apparatus, in which DNA polymerase alpha and primase are two key enzymatic components. Primase, which is a heterodimer of a small subunit and a large subunit, synthesizes small RNA primers for the Okazaki fragments made during discontinuous DNA replication. The protein encoded by this gene is the small, 49 kDa primase subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRIM1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PRIM1
Gene Accession No. P20664, P49642
Gene Alias AI324982; DNA primase 1; DNA primase 49 kDa subunit; DNA primase small subunit; DNA primase subunit 48; DNA primase, p49 subunit; MGC12308; p49; PRI1; PRIM1; primase (DNA) subunit 1; primase p49 subunit; primase polypeptide 1, 49kDa; primase, DNA, polypeptide 1; primase, DNA, polypeptide 1 (49kDa)
Gene Symbols PRIM1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 121-420 of human PRIM1 (NP_0009371).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19075, 5557
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.