Learn More
Invitrogen™ PRIM2 Monoclonal Antibody (4G2)

Description
Peptide Sequence: MEFSGRKWRK LRLAGDQRNA SYPHCLQFYL QPPSENISLI EFENLAIDRV KLLKSVENLG VSYVKGTEQY QSKLESELRK LKFSYRENLE DEYEPRRRDH ISHFILRLAY CQSEELRRWF IQQEMDLLRF RFSILPKDKI QDFLKDSQLQ FEAVSIFL.
Specifications
Specifications
| Antigen | PRIM2 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 4G2 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | Prim2 |
| Gene Accession No. | P49643 |
| Gene Alias | AI323589; DNA polymerase alpha subunit III (primase); DNA primase 58 kDa subunit; DNA primase large subunit; DNA primase, p58 subunit; p58; Pola3; PRIM2; PRIM2A; primase (DNA) subunit 2; primase, DNA, polypeptide 2; primase, DNA, polypeptide 2 (58kDa) |
| Gene Symbols | Prim2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.