Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRIM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588189
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588189 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588189 Supplier Invitrogen™ Supplier No. PA588189
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEFSGRKWRK LRLAGDQRNA SYPHCLQFYL QPPSENISLI EFENLAIDRV KLLKSVENLG VSYVKGTEQY QSKLESELRK LKFSYRENLE DEYEPRRRDH ISHFILRLAY CQSEELRRWF IQQEMDLLRF RFSILPKDKI QDFLKDSQLQ FEAISDEEKT LREQEIVASS; Positive Samples: A375, H460, HeLa, HT-29, Mouse spleen.

DNA primase is the polymerase that synthesizes small RNA primers for the Okazaki fragments made during discontinuous DNA replication.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRIM2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.67 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Prim2
Gene Accession No. P33610, P49643
Gene Alias AI323589; DNA polymerase alpha subunit III (primase); DNA primase 58 kDa subunit; DNA primase large subunit; DNA primase, p58 subunit; p58; Pola3; PRIM2; PRIM2A; primase (DNA) subunit 2; primase, DNA, polypeptide 2; primase, DNA, polypeptide 2 (58kDa)
Gene Symbols Prim2
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19076, 5558
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.