Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRIMA1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563809
Description
Immunogen sequence: KRKPLRKDEN GTSVAEYPMS ASQSNKGVDV NNAVV Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 89%.
Required to anchor acetylcholinesterase (ACHE) to the basal lamina of the neuromuscular junction and to the membrane of neuronal synapses in brain. Also able to organize ACHE into tetramers.
Specifications
PRIMA1 | |
Polyclonal | |
Unconjugated | |
PRIMA1 | |
acetylcholinesterase membrane anchor precursor PRiMA; PRIMA; PRIMA1; proline rich membrane anchor 1; proline-rich membrane anchor 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
145270 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86XR5 | |
PRIMA1 | |
Recombinant protein corresponding to Human PRIMA1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction