Learn More
Description
- Hoods with collar and full bib have an assigned protection factor (APF) of 1000
- Manifold directs air over lens to reduce fogging
- PA121 and PA141 are coated for protection from intermittent splash
- Primair 100 Series hoods for use with Compact Air 200 Series PAPR and CF1000 Series continuous flow airline feature a comfortable headgear that directs air over the lens for cooling comfort
- Triple adjustment headgear for a comfortable fit
Specifications
Specifications
| Color | White |
| Product Type | Respirator Hood |
| For Use With (Equipment) | Compact Air 200 Series PAPR, CF1000 Series continuous flow airline |
| Includes | Neck seal |
| Model | PA202 |
| Quantity | 10/Cs. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.