Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Learning Resources Primary Science™ Jumbo Eyedroppers with Stand

Click to view available options
Class Size:
1 Class
Description
- Have fun experimenting with cause and effect while building fine motor skills at the same time.
- This set of 6 perfectly sized and colorful droppers store vertically in a convenient stand.
- A great addition to any science center or sensory table.
Specifications
Specifications
Grade Level | Elementary, Intermediate - Middle School, High School |
Grade | K to 12 |
Class Size | 1 Class |
Product Type | Eyedropper Set |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction