Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Learning Resources Primary Science™ Jumbo Magnifiers, Set of 12 in Display (without stand)

Click to view available options
Class Size:
12 Students
Description
- Introduce early scientific exploration and investigation with these extra-large magnifiers that are sized perfectly for small hands.
- Great for taking a closer look at plants, animals and insects in life science lessons.
- Built-in stands allow hands-free viewing.
- Ages 3+.
- Set of 12.
Specifications
Specifications
| Grade Level | Elementary, Intermediate - Middle School, High School |
| Grade | K to 12 |
| Class Size | 12 Students |
| Product Type | Magnifier |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction