Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Monkey IL-6 ELISA Kit (Colorimetric)

Click to view available options
Quantity:
96 Tests
Description
The Primate IL-6 ELISA Kit (Colorimetric) measures primate IL-6 in cell culture supernatant, serum (baboon, cynomolgus).
Specifications
Specifications
Assay Range | 1.9 to 120 U/mL |
Assay Sensitivity | 1.3 U/mL |
Product Type | ELISA Kit (Colorimetric) |
Sample Type | Cell Culture Supernatant, Cynomolgus Serum, Serum (baboon, cynomolgus) |
Specificity | The interference of circulating factors of the immune systemwas evaluated by spiking these proteins at physiologically relevant concentrations into a monkey IL-6 positive serum. There was no crossreactivity detected. |
Target Species | Primate |
For Use With (Application) | ELISA |
Gene ID (Entrez) | 3569 |
Gene Symbol | IL6 |
Kit Contents | Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to monkey IL-6, Biotin-Conjugate anti-monkey IL-6 monoclonal antibody, Streptavidin-HRP, Monkey IL-6 Standard lyophilized, 240U/mL upon reconstitution, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction