Learn More
Applied Biosystems™ Primer Express™ Software v3.0.1 License
Description
Easy-to-Learn Software
The Primer Express™ Software v3.0.1 is flexible, easy to learn and to use, and requires minimal optimization. It includes an algorithm optimized specifically for use with TaqMan™-based reagents, recommended primer⁄probe concentrations, and default thermal cycling conditions. Primer Express™ Software also supports assays based on the SYBR™ Green I dye. This minimizes the need for assay optimization and streamlines the design process.
The software also allows customized designs if the probe and⁄or one or more primers are known. Default and adjustable parameters (Figure 1) provide the flexibility required for all users, regard
Simple to Use
Simplicity and flexibility are the hallmarks of Primer Express™ Software v3.0.1, which is fully compatible with all StepOne™, StepOnePlus™, 7300, 7500, 7500 Fast, 7900HT, ViiA™ 7, and QuantStudio™ real-time PCR systems. Whether your research includes gene quantitation or allelic discrimination studies, Primer Express™ Software belongs in your lab.
Automated and Manual Primer⁄Probe Design
The Primer Express™ Software v3.0.1 will design your primers and probes automatically, or you can design them manually. For automated design, simply choose either TaqMan ™ or TaqMan™ MGB assays. If you know the sequence, you need only to import (or copy and paste) the sequence and quickly view your results. Alternatively, if you prefer to design the primers and probes manually, the procedure is equally simple and straight-forward as well.
Rapid Assay Development Guidelines
The Primer Express ™ Software v3.0.1 was designed with our Rapid Assay Development Guidelines (RADG). The RADG are a simple yet comprehensive set of guidelines for use with our real-time PCR instruments and reagents. Many traditional variables, including magnesium chloride concentration and the thermal cycling protocol itself, have been standardized, which substantially reduces assay development time.
For Research Use Only. Not for use in diagnostics procedures.
Specifications
Specifications
| Content And Storage | Store at Room Temperature, Included w/ StepOne, StepOnePlus, 7300, 7500, 7500 Fast, 7900HT, ViiA 7, QuantStudio 6, QuantStudio 7, and QuantStudio 12k Flex, Runs computers on Windows 10 OS for 7500, 7500 Fast, QuantStudio 6, QuantStudio 7, and QuantStudio |
| For Use With (Equipment) | 7300 System, 7500 Fast System, 7500 System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, StepOne™, StepOnePlus™, ViiA™ 7 System |
| Product Line | Primer Express |
| Software Type | Primer Design |
| Product Type | License |
| For Use With (Application) | Real Time PCR (qPCR) |
| Operating System | Windows 10 |
| Quantity | 1 user |
Frequently Asked Questions (FAQs)
You can directly see all the primer and probe sequences from a test design right away using the Primer Express Software. Open the software and choose File => New. Choose either TaqMan MGB Quantification or TaqMan Quantification for Gene Expression Assay design. Copy/paste your gene sequence of interest into the new window and click on the green triangle to get primer/probe designs.
Yes, you can use Primer Express v3.0.1 on Win10 computers.
Primer and probe sets that satisfy the optimal length and Tm requirements are in red. Sets that don't satisfy one or the other or both of these parameters are in blue or green.
The Primer Express 2.0 software was developed to work on Windows 2000 operating systems. The "not enough memory" error message is generated due to incompatibility with the newer processor in the computers and the algorithm of the Primer Express v2.0 software. We have developed a patch to help over come this error message. You can download this patch from our website by going to the software downloads option under the Technical Resources page on our website. Click on ABI PRISM 7000 Sequence Detection System, then click on the PC link next to Primer Express P4 Enabler. Also download the installation instructions by clicking on the Read Me link.
This error message can occur while attempting to install the Primer Express software, or sometimes when launching the software after it has been installed on the computer. To create a new archive file, hold down Alt key (to the left of the space bar) while opening the Primer Express application. You should get the browser window shown below. Click on New to create a New Archive.
The name PXArchive will automatically populate in the file name field. You may keep that name or change it to something different. Once you've named the Archive file, click on save. You should now be able to continue with the installation or use of the Primer Express Software.
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.