Learn More
Thermo Scientific™ Printers for CryoMed™ and CryoPlus™ Freezers
GSA_VA
Catalog No. tf4000566
Click to view available options
Type:
Thermal Data Printer
Thermal Printer Paper
Voltage:
120 V
Not Applicable
Enhance the functionality for Thermo Scientific™ CryoMed™ controlled-rate freezers with this optional printer.
Enhance the functionality for Thermo Scientific™ CryoMed™ controlled-rate freezers with Thermo Scientific™ Thermal Printers for CryoMed and CryoPlus™ Freezers. These printers are available in 120V or 220V models, and include cable assembly and RS-232 connector kit.
  • Model 4000565 - 120V, 60Hz printer with cable assembly and RS-232 connector kit
  • Model 4000665 - 220V, 50Hz printer with cable assembly and RS-232 connector kit
  • Model 4000566 - Thermal printer paper (5 rolls/package)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Type Thermal Printer Paper
For Use With (Equipment) All CryoMed Controlled-Rate Freezers
Voltage Not Applicable
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.