Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Cable Lockout Device
Click to view available options
Includes:
10 ft. cable
6 ft. cable
Description
- Versatile device for all of your unusual energy isolating devices that are difficult to lock out
- 6 lock holes in the handle are 9/32 in. in diameter, which accommodates most lock shackles and hasps
- Includes 10 in. cable
Specifications
Specifications
Product Type | Tab-cinching Cable Lockout Device |
Color | Red |
Material | Plastic |
Dimensions (L x W x H) | 0.65 x 5.84 x 2 in. |
Includes | 6 ft. cable |
Quantity | 1 Ea. |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction