Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Large White Lockout Center

Catalog No. 19104058 Shop All Brady Corporation Products
Click to view available options
Color:
White

Develop a safety plan to protect workers and the worksite using lockout products. The Brady™ Large White Lockout Center is the perfect solution to get started on a successful lockout program by organizing and storing padlocks.

  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need

Specifications

Color White
Dimensions (L x W) 23.75 x 30 in.
Displayed Text SAFETY LOCKOUT/TAGOUT CENTER USE LOCKS, TAGS AND HASPS TO DISABLE EQUIPMENT PRIOR TO MAINTENANCE, CLEANING OR INSPECTION
Height (English) 30 in.
Includes (2) danger label; (2) lockout hasp; (8) assorted kd safety padlock; 1 to 2.5 in gate valve lockout; 110 v plug lockout; 3-in-1 plug valve lockout; lockout tag; small ball valve lockout
Quantity 1 Kit
Width (English) 23.75 in.

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.