Learn More
United Scientific Supplies Prism and Lens Sets
SDPSCI_ED
Catalog No. p-3071315
Click to view available options
Material:
Acrylic
Glass
Teach a wide variety of optical principles.
Set housed in a wooden case. Shipping weight: 2 lb. (0.9kg); box dimensions: 9.25 x 8 x 2 in. (23.1 x 20 x 5cm). Glass and acrylic sets each have seven components.
  • Double convex lens
  • Double concave lens (can be used together with double convex lens)
  • Semicircular lens
  • Trapezoidal prism
  • Small right-angle prism
  • Equilateral prism
  • Large right-angle refraction prism
Specifications
Material Acrylic
For Use With (Application) Teach a wide variety of optical principles
Product Type Prism and Lens Set
Includes Double convex lens, double concave lens, semicircular lens, trapezoidal prism, small right angle prism, equilateral prism, large right angle refraction prism
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.