Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRK1/PRK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106263
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106263 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106263 Supplier Invitrogen™ Supplier No. PA5106263
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PRK1/2.

Receptor-like kinase involved in the control of pollen germination and pollen tube polar growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRK1/PRK2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PKN1
Gene Accession No. O08874, P70268, Q16512, Q16513, Q63433, Q8BWW9
Gene Alias 6030436C20Rik; AI507382; cardiolipin-activated protein kinase Pak2; DBK; F730027O18Rik; p140 kinase; PAK1; PAK-1; PAK2; Pak-2; Pkn; PKN gamma; Pkn1; Pkn2; PKN-ALPHA; Prk1; Prk2; Prkcl1; PRKCL2; PRO2042; protease-activated kinase 1; protease-activated kinase 2; protein kinase C-like 1; Protein kinase C-like 2; protein kinase C-like PKN; protein kinase C-related kinase 1; protein kinase N1; protein kinase N2; protein kinase PKN; protein kinase PKN-alpha; Protein-kinase C-related kinase 1; protein-kinase C-related kinase 2; serine/threonine kinase 3; serine/threonine kinase 7; serine/threonine protein kinase N; serine/threonine-protein kinase N1; Serine/threonine-protein kinase N2; serine-threonine kinase N; serine-threonine protein kinase N; Stk3; Stk7
Gene Symbols PKN1, PKN2
Host Species Rabbit
Immunogen A synthesized peptide derived from human PKN1(Accession Q16512), corresponding to amino acid residues L748-W798.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109333, 207122, 29355, 320795, 5585, 5586
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.