Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRKAA1 (phospho S487) Rabbit anti-Human, Polyclonal, Abnova™

Catalog No. 89033725 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-033-725 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-033-725 Supplier Abnova Corporation Supplier No. PAB30166

Rabbit Polyclonal Antibody

Rabbit polyclonal antibody raised against partial synthetic phosphopeptide of human PRKAA1.

  • High specificity
  • High reactivity

Specifications

Antigen PRKAA1 (phospho S487)
Applications Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial synthetic phosphopeptide of human PRKAA1.
Dilution Immunofluorescence (1:100-1:200) Immunohistochemistry (1:50-1:100) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation In 1.0 mg/mL PBS (without Mg2+ and Ca2+), 150 mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide).
Gene Accession No. Q13131
Gene Alias AMPK/AMPKa1/MGC33776/MGC57364
Gene Symbols PRKAA1
Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to residues surrounding S487 of human PRKAA1.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5562
Target Species Human
Content And Storage Store at 4°C for short term use. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.