Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRKAB2 Mouse anti-Human, Clone:8F5B2, Abnova™

Catalog No. 89334389 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-389 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-389 Supplier Abnova Corporation Supplier No. MAB17941
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant human PRKAB2.

The protein encoded by this gene is a regulatory subunit of the AMP-activated protein kinase (AMPK). AMPK is a heterotrimer consisting of an alpha catalytic subunit, and non-catalytic beta and gamma subunits. AMPK is an important energy-sensing enzyme that monitors cellular energy status. In response to cellular metabolic stresses, AMPK is activated, and thus phosphorylates and inactivates acetyl-CoA carboxylase (ACC) and beta-hydroxy beta-methylglutaryl-CoA reductase (HMGCR), key enzymes involved in regulating de novo biosynthesis of fatty acid and cholesterol. This subunit may be a positive regulator of AMPK activity. It is highly expressed in skeletal muscle and thus may have tissue-specific roles. [provided by RefSeq]

Specifications

Antigen PRKAB2
Applications ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 8F5B2
Conjugate Unconjugated
Description protein kinase, AMP-activated, beta 2 non-catalytic subunit
Dilution ELISA (1:10000)
Flow Cytometry (1:200-1:400)
Immunofluorescence (1:200-1:1000)
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections)
Western Blot (1:100-1:500)
The optimal working dilution should be determined by the end user
Formulation In PBS (0.05% sodium azide)
Gene PRKAB2
Gene Alias MGC61468
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-120 of human PRKAB2.
Quantity 100 μg
Gene ID (Entrez) 5565
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.