Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRKACG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112671
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112671 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112671 Supplier Invitrogen™ Supplier No. PA5112671
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PRKACG Polyclonal Antibody for Western Blot, IHC (P), ELISA

PRKACG (PRKAC gamma) is a third isoform of the catalytic subunit of cAMP-dependent protein kinase. It was isolated from a human testis cDNA library and was clearly derived from a gene distinct from C-alpha and C-beta and showed tissue-specific expression. Whereas at the amino acid level C-alpha and C-beta showed 93% homology, C-gamma showed only about 80% homology to both C-alpha and C-beta. The PRKACG gene is intronless, contains remnants of a poly(A) tail, is flanked by direct repeats, and is co-linear with the PRKACA gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRKACG
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PRKACG
Gene Accession No. P22612
Gene Alias BDPLT19; cAMP-dependent protein kinase catalytic subunit gamma; KAPG; PKA C-gamma; PKACg; PRKACG; protein kinase cAMP-activated catalytic subunit gamma; protein kinase, cAMP-dependent, catalytic, gamma; protein kinase, cAMP-dependent, gamma catalytic subunit; serine(threonine) protein kinase
Gene Symbols PRKACG
Host Species Rabbit
Immunogen Recombinant Human cAMP-dependent protein kinase catalytic subunit gamma protein (1-43AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5568
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.