Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRKAG2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NBP189324
Description
PRKAG2 Polyclonal specifically detects PRKAG2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
PRKAG2 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
AAKG, AAKG2, CMH6AMPK subunit gamma-2, H91620pAMPK gamma2, protein kinase, AMP-activated, gamma 2 non-catalytic subunit, WPWS5'-AMP-activated protein kinase subunit gamma-2 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of human PRKAG2 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
PRKAG2 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:STPTQVTKQHTFPLESYKHEPERLENRIYASSSPPDTGQRFCPSSFQSPTRPPLASPTHYAPSKAAALAAALGPAEAGMLEKLEFEDEAVEDSESGVYMRFMRSHK | |
0.1 mL | |
Lipid and Metabolism | |
51422 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction