Learn More
Invitrogen™ PRKAR1A Polyclonal Antibody

Description
PRKAR1A Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | PRKAR1A |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | PRKAR1A |
| Gene Accession No. | P10644 |
| Gene Alias | 1300018C22Rik; ACRDYS1; ADOHR; cAMP-dependent protein kinase regulatory subunit RIalpha; cAMP-dependent protein kinase type I-alpha regulatory chain; cAMP-dependent protein kinase type I-alpha regulatory subunit; cAMP-dependent protein kinase type I-alpha regulatory subunit, N-terminally processed; CAR; Carney complex type 1; CNC; CNC1; PKR1; PPNAD1; PRKAR1; Prkar1a; protein kinase A type 1a regulatory subunit; protein kinase cAMP-dependent type I regulatory subunit alpha; protein kinase, cAMP dependent regulatory, type 1, alpha; protein kinase, cAMP dependent regulatory, type I, alpha; protein kinase, cAMP-dependent, regulatory subunit type I alpha; protein kinase, cAMP-dependent, regulatory, type I, alpha; RIalpha; RIIA; Tissue-specific extinguisher 1; Tse1; Tse-1 |
| Gene Symbols | PRKAR1A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.