Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRKAR1A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580936
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human PRKAR1A/PRKAR1/PKR1.
cAMP is a signaling molecule important for a variety of cellular functions. cAMP exerts its effects by activating the cAMP-dependent protein kinase, which transduces the signal through phosphorylation of different target proteins. The inactive kinase holoenzyme is a tetramer composed of two regulatory and two catalytic subunits. cAMP causes the dissociation of the inactive holoenzyme into a dimer of regulatory subunits bound to four cAMP and two free monomeric catalytic subunits. Four different regulatory subunits and three catalytic subunits have been identified in humans. This gene encodes one of the regulatory subunits. This protein was found to be a tissue-specific extinguisher that down-regulates the expression of seven liver genes in hepatoma x fibroblast hybrids. Mutations in this gene cause Carney complex (CNC). This gene can fuse to the RET protooncogene by gene rearrangement and form the thyroid tumor-specific chimeric oncogene known as PTC2. A nonconventional nuclear localization sequence (NLS) has been found for this protein which suggests a role in DNA replication via the protein serving as a nuclear transport protein for the second subunit of the Replication Factor C (RFC40). Three alternatively spliced transcript variants encoding the same protein have been observed.
Specifications
PRKAR1A | |
Polyclonal | |
Unconjugated | |
PRKAR1A | |
1300018C22Rik; ACRDYS1; ADOHR; cAMP-dependent protein kinase regulatory subunit RIalpha; cAMP-dependent protein kinase type I-alpha regulatory chain; cAMP-dependent protein kinase type I-alpha regulatory subunit; cAMP-dependent protein kinase type I-alpha regulatory subunit, N-terminally processed; CAR; Carney complex type 1; CNC; CNC1; PKR1; PPNAD1; PRKAR1; Prkar1a; protein kinase A type 1a regulatory subunit; protein kinase cAMP-dependent type I regulatory subunit alpha; protein kinase, cAMP dependent regulatory, type 1, alpha; protein kinase, cAMP dependent regulatory, type I, alpha; protein kinase, cAMP-dependent, regulatory subunit type I alpha; protein kinase, cAMP-dependent, regulatory, type I, alpha; RIalpha; RIIA; Tissue-specific extinguisher 1; Tse1; Tse-1 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
5573 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with no preservative | |
P10644 | |
PRKAR1A | |
Recombinant Human PRKAR1A/PRKAR1/PKR1 Protein (Met1-Val381). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction