Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRKAR1A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501732845 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-2845 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-2845 Supplier Proteintech Group Inc Supplier No. 203581AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The second messenger cyclic AMP (cAMP) mediates diverse cellular responses to external signals such as proliferation, ion transport, regulation of metabolism and gene transcription by activation of the cAMP-dependent protein kinase (cAPK or PKA). Activation of PKA occurs when cAMP binds to the two regulatory subunits of the tetrameric PKA holoenzyme, resulting in release of active catalytic subunits. Four different PKA regulatory subunits have been identified, designated Ia, Ib, IIa and IIb. The PKA Ia reg protein is a tissue-specific extinguisher that downregulates the expression of seven liver genes in hepatoma x fibroblast hybrids. Functional null mutations in the gene that codes for PKA Ia reg cause Carney complex (CNC). CNC is an autosomal domit multiple neoplasia syndrome. CNC is associated with a variety of characterized symptoms such as cardiac and other myxomas, spotty skin pigmentation, endocrine tumors and psammomatous melanotic schwannomas.

Specifications

Antigen PRKAR1A
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.62 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PRKAR1A
Gene Accession No. P09456, P10644, Q9DBC7
Gene Alias CAR, CNC, CNC1, PKR1, PPNAD1, PRKAR1, PRKAR1A, Tissue specific extinguisher 1, TSE1
Gene Symbols Prkar1a
Host Species Rabbit
Immunogen PRKAR1A Fusion Protein Ag14216
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19084, 25725, 5573
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.