Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRKAR1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587652
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587652 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587652 Supplier Invitrogen™ Supplier No. PA587652
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WVTNISEGGS FGELALIYGT PRAATVKAKT DLKLWGIDRD SYRRILMGST LRKRKMYEEF LSKVSILESL EKWERLTVAD ALEPVQFEDG EKIVVQGEPG D; Positive Samples: U-251MG, HeLa, Jurkat, HepG2, BT-474, Mouse brain, Mouse lung, Rat brain, Rat testis; Cellular Location: Cell membrane.

The second messenger cyclic AMP (cAMP) mediates diverse cellular responses to external signals such as proliferation, ion transport, regulation of metabolism and gene transcription by activation of the cAMP-dependent protein kinase (cAPK or PKA). Activation of PKA occurs when cAMP binds to the two regulatory subunits of the tetrameric PKA holoenzyme resulting in release of active catalytic subunits. Three catalytic (C) subunits have been identified that each represent specific gene products. Activation of transcription upon elevation of cAMP levels results from translocation of PKA to the nucleus where it phosphorylates the transcrip- tion factor cAMP response element binding protein (CREB) on serine 133 which in turn leads to TFIIB binding to TATA-box-binding protein TBP1, thus linking phospho-CREB to the pol II transcription initiation complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRKAR1B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PRKAR1B
Gene Accession No. P12849, P31321, P81377
Gene Alias AI385716; cAMP dependent protein kinase, subunit type 1 beta; cAMP dependent regulatory type I beta subunit transcript 2; cAMP dependent regulatory type I beta subunit transcript 3; cAMP dependent regulatory type I beta subunit transcript 4; cAMP-dependent protein kinase type I-beta regulatory subunit; PRKAR1; PRKAR1B; protein kinase cAMP-dependent type I regulatory subunit beta; protein kinase, cAMP dependent regulatory, type I beta; protein kinase, cAMP dependent regulatory, type I, beta; protein kinase, cAMP-dependent, regulatory subunit type I beta; protein kinase, cAMP-dependent, regulatory, type I, beta; RGD1563094; RIbeta
Gene Symbols PRKAR1B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 190-290 of human PRKAR1B (NP_0027261).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19085, 25521, 5575
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.