Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRKAR2A Monoclonal Antibody (OTI4A3), TrueMAB™, OriGene
SDP

Catalog No. 502542503
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2503 30 μL
50-167-2285 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2503 Supplier OriGene Supplier No. TA501144S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PRKAR2A Monoclonal antibody specifically detects PRKAR2A in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen PRKAR2A
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4A3
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PRKAR2A
Gene Accession No. P13861
Gene Alias 1110061A24Rik; AI317181; AI836829; cAMP-dependent protein kinase regulatory subunit RII alpha; cAMP-dependent protein kinase type II-alpha regulatory subunit; cAMP-dependent protein kinase, regulatory subunit alpha 2; MGC3606; PKR2; PRKAR2; Prkar2a; protein kinase A, RII-alpha subunit; protein kinase cAMP-dependent regulatory type II alpha; protein kinase cAMP-dependent type II regulatory subunit alpha; protein kinase, cAMP dependent regulatory, type II alpha; protein kinase, cAMP-dependent, regulatory subunit type II alpha; protein kinase, cAMP-dependent, regulatory, type 2, alpha; protein kinase, cAMP-dependent, regulatory, type II, alpha; RII(alpha); unnamed protein product
Host Species Mouse
Immunogen Full length human recombinant protein of human PRKAR2A produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5576
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.