Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRKAR2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590367
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590367 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590367 Supplier Invitrogen™ Supplier No. PA590367
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSHIQIPPGL TELLQGYTVE VLRQQPPDLV EFAVEYFTRL REARAPASVL PAATPRQSLG HPPPEPGPDR VADAKGDSES EEDEDLEVPV PSRFNRRVSV CAETYNPDEE EEDTDPRVIH PKTDEQRCRL QEACKDILLF KNLDQEQLSQ VLDAMFERIV KADEHVIDQG DDGDNFYVIE RGTYDILVTK DNQTRSVGQY DNRGSFGELA LMYNTPRAAT IVATSEGSLW GLDRVTFRRI IVKNNAKKRK MFESFIESVP LLKSLEVSER MKIVDVIGEK IYKDGERIIT QGEKADSFYI IESGEVSILI RSRTKSNKDG GNQEVEIARC HKGQYFGELA LVTNKPRAAS AYAVGDVKCL VMDVQAFERL LGPCMDIMKR NISHYEEQLV KMFGSSVDLG NLGQ; Positive Samples: HeLa, Jurkat, SW480, LO2, Mouse lung, Rat skeletal muscle; Cellular Location: Cell membrane, Cytoplasm.

cAMP is a signaling molecule important for a variety of cellular functions. cAMP exerts its effects by activating the cAMP-dependent protein kinase (AMPK), which transduces the signal through phosphorylation of different target proteins. The inactive holoenzyme of AMPK is a tetramer composed of two regulatory and two catalytic subunits. cAMP causes the dissociation of the inactive holoenzyme into a dimer of regulatory subunits bound to four cAMP and two free monomeric catalytic subunits. Four different regulatory subunits and three catalytic subunits of AMPK have been identified in humans. The protein encoded by this gene is one of the regulatory subunits. This subunit can be phosphorylated by the activated catalytic subunit. It may interact with various A-kinase anchoring proteins and determine the subcellular localization of AMPK.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRKAR2A
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 4.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene PRKAR2A
Gene Accession No. P12367, P12368, P13861
Gene Alias 1110061A24Rik; AI317181; AI836829; cAMP-dependent protein kinase regulatory subunit RII alpha; cAMP-dependent protein kinase type II-alpha regulatory subunit; cAMP-dependent protein kinase, regulatory subunit alpha 2; MGC3606; PKR2; PRKAR2; Prkar2a; protein kinase A, RII-alpha subunit; protein kinase cAMP-dependent regulatory type II alpha; protein kinase cAMP-dependent type II regulatory subunit alpha; protein kinase, cAMP dependent regulatory, type II alpha; protein kinase, cAMP-dependent, regulatory subunit type II alpha; protein kinase, cAMP-dependent, regulatory, type 2, alpha; protein kinase, cAMP-dependent, regulatory, type II, alpha; RII(alpha); unnamed protein product
Gene Symbols PRKAR2A
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19087, 29699, 5576
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.