Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRKAR2B Polyclonal Antibody

PRKAR2B Polyclonal Antibody

Supplier:  Thermo Scientific PA528266

Catalog No. PIPA528266


Only null left
Add to Cart

Description

Description

PRKAR2B Polyclonal Antibody for Western Blot, ICC/IF, IHC (F), IHC (P)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

PRKAR2B
Polyclonal
Unconjugated
Prkar2b
AI451071; AW061005; cAMP-dependent protein kinase RII-beta regulatory subunit; cAMP-dependent protein kinase type II-beta regulatory chain; cAMP-dependent protein kinase type II-beta regulatory subunit; H_RG363E19.2; Pkarb2; PKARIIbeta; PRKAR2; PRKAR2B; protein kinase cAMP-dependent type II regulatory subunit beta; protein kinase, cAMP dependent regulatory, type II beta; protein kinase, cAMP-dependent, regulatory subunit type II beta; protein kinase, cAMP-dependent, regulatory, type II, beta; RATDNA; RII(beta); RII-BETA; Type II beta regulatory subunit of cAMP-dependent protein kinase; WUGSC:H_RG363E19.2
Rabbit
Antigen Affinity Chromatography
RUO
19088, 24679, 5577
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.67 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P12369, P31323, P31324
Prkar2b
Recombinant fragment corresponding to a region within amino acids 177 and 391 of Human PKA 2 beta.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.