Learn More
Invitrogen™ PRKAR2B Polyclonal Antibody

Description
PRKAR2B Polyclonal Antibody for Western Blot, IHC (P), IP
Specifications
Specifications
| Antigen | PRKAR2B |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.03% ProClin 300; pH 7 |
| Gene | Prkar2b |
| Gene Accession No. | P31323 |
| Gene Alias | AI451071; AW061005; cAMP-dependent protein kinase RII-beta regulatory subunit; cAMP-dependent protein kinase type II-beta regulatory chain; cAMP-dependent protein kinase type II-beta regulatory subunit; H_RG363E19.2; Pkarb2; PKARIIbeta; PRKAR2; PRKAR2B; protein kinase cAMP-dependent type II regulatory subunit beta; protein kinase, cAMP dependent regulatory, type II beta; protein kinase, cAMP-dependent, regulatory subunit type II beta; protein kinase, cAMP-dependent, regulatory, type II, beta; RATDNA; RII(beta); RII-BETA; Type II beta regulatory subunit of cAMP-dependent protein kinase; WUGSC:H_RG363E19.2 |
| Gene Symbols | Prkar2b |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.