Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRKAR2B Recombinant Rabbit Monoclonal Antibody (JA11-69)
GREENER_CHOICE

Catalog No. PIMA532662
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532662 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532662 Supplier Invitrogen™ Supplier No. MA532662
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

cAMP is a signaling molecule important for a variety of cellular functions. cAMP exerts its effects by activating the cAMP-dependent protein kinase, which transduces the signal through phosphorylation of different target proteins. The inactive kinase holoenzyme is a tetramer composed of two regulatory and two catalytic subunits. cAMP causes the dissociation of the inactive holoenzyme into a dimer of regulatory subunits bound to four cAMP and two free monomeric catalytic subunits. Four different regulatory subunits and three catalytic subunits have been identified in humans. The protein encoded by this gene is one of the regulatory subunits. This subunit can be phosphorylated by the activated catalytic subunit. This subunit has been shown to interact with and suppress the transcriptional activity of the cAMP responsive element binding protein 1 (CREB1) in activated T cells. Knockout studies in mice suggest that this subunit may play an important role in regulating energy balance and adiposity. The studies also suggest that this subunit may mediate the gene induction and cataleptic behavior induced by haloperidol.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRKAR2B
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Recombinant Monoclonal
Clone JA11-69
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene Prkar2b
Gene Accession No. P12369, P31323, P31324
Gene Alias AI451071; AW061005; cAMP-dependent protein kinase RII-beta regulatory subunit; cAMP-dependent protein kinase type II-beta regulatory chain; cAMP-dependent protein kinase type II-beta regulatory subunit; H_RG363E19.2; Pkarb2; PKARIIbeta; PRKAR2; PRKAR2B; protein kinase cAMP-dependent type II regulatory subunit beta; protein kinase, cAMP dependent regulatory, type II beta; protein kinase, cAMP-dependent, regulatory subunit type II beta; protein kinase, cAMP-dependent, regulatory, type II, beta; RATDNA; RII(beta); RII-BETA; Type II beta regulatory subunit of cAMP-dependent protein kinase; WUGSC:H_RG363E19.2
Gene Symbols Prkar2b
Host Species Rabbit
Immunogen Synthetic peptide within Human PKA 2 beta aa 369-418.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19088, 24679, 5577
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.