Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRKCB, Rabbit anti-Human, Clone: N11-S, Abnova™
Rabbit Monoclonal Antibody
Supplier: Abnova Corporation MAB15891
Specifications
PRKCB | |
Monoclonal | |
Unconjugated | |
In 20 mM Tris-HCl buffer, pH 8.0 (10 mg/mL BSA, 0.05% Sodium Azide). | |
MGC41878/PKC-beta/PKCB/PRKCB1/PRKCB2 | |
Rabbit | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
N11-S | |
ELISA (1:20000-1:100000) Immunocytochemistry Immunoprecipitation Western Blot (1:10000) The optimal working dilution should be determined by the end user. | |
protein kinase C, beta | |
PRKCB | |
A synthetic peptide corresponding to amino acids 663-672 at C-terminus of human PRKCB2. | |
RUO | |
5579 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction