Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRKCBP1 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP119131UV
Description
PRKCBP1 Polyclonal antibody specifically detects PRKCBP1 in Human samples. It is validated for Western BlotSpecifications
PRKCBP1 | |
Polyclonal | |
Western Blot | |
ESP-2, HED-2, zyxin, Zyxin-2 | |
The immunogen recognized by this antibody maps to a region between residue 1136 and 1186 of human protein kinase C binding protein 1 (zinc finger, MYND-type containing 8) using the numbering given in entry CAI21842.1 (GeneID 23613). | |
0.1 mL | |
Signal Transduction | |
23613 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction