Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRKCDBP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP25789725UL
Description
PRKCDBP Polyclonal specifically detects PRKCDBP in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
PRKCDBP | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
Cavin-3, hSRBC, MGC20400, protein kinase C, delta binding protein, sdr-related gene product that binds to c-kinase, Serum deprivation response factor-related gene product that binds to C-kinase, SRBCprotein kinase C delta-binding protein | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
PRKCDBP | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:EAQPEAQPALEPTLEPEPPQDTEEDPGRPGAAEEALLQMESVA | |
25 μL | |
Diabetes Research, Lipid and Metabolism | |
112464 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction