Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRKCDBP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531411
Description
Recommended positive controls: HeLa. Predicted reactivity: Mouse (88%), Rat (90%), Bovine (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
JAB-1 is a 38 kDa protein which was originally described as a coactivator for cJUN. It is present in a large protein complex called the Signalosome , which has been proposed to play a signaling role in mammalian cells. This protein complex appears to be analogous to the COP9 complex in plants, which is important in light-mediated plant development. Although the specific function of JAB-1 in this complex is unclear, it has recently been implicated in targeting proteins for degradation by the proteasome.
Specifications
PRKCDBP | |
Polyclonal | |
Unconjugated | |
CAVIN3 | |
3110015B12Rik; 6330514M23Rik; Caveolae-associated protein 3; cavin 3; CAVIN3; cavin-3; D3T-inducible gene 2 protein; Dig2; DIG-2; Dithiolethione-inducible gene 2 protein; dithiolethione-inducible gene-2; hSRBC; MGC20400; PKC-delta binding protein; Prkcdbp; protein kinase C delta binding protein; protein kinase C delta-binding protein; protein kinase C, delta binding protein; sdr-elated gene product that binds to c-kinase; sdr-related gene product that binds to c-kinase; serum deprivation response factor-related gene product that binds to C-kinase; SRBC | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
112464 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q969G5 | |
CAVIN3 | |
Recombinant fragment corresponding to a region within amino acids 1 and 218 of Human PRKCDBP. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction